Cellulose for preparative chromatography
Catalog # 1.02331.0500
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Velikost částic:10 - 160 µm
- Bal.:500 g
- Popis:Cellulose micro-crystalline Avicel®
Specifications
About this item
Micro-crystalline cellulose is a hydrophilic polysaccharide packing material preferred for the raw separation of amino acids and related compounds. Cellulose is also used for the gentle purification of bio-molecules. Because of the organic nature of cellulose, it can only be used in pre-swollen state and under gentle or hydrostatic pressure.