Stopcock, three way, BD Connecta™
Catalog # BDAM394501
Supplier: BD Medical
Specifications
- Popis:Stopcock, three way, with BD Q-Syte™ luer access split septum device
- Bal.:50
Specifications
About this item
This three way stopcock has a flow rate of 23,4 L/hr and a prime volume of 0,31 ml.