Ammonium molybdate reagent for silica determination using Lovibond₂ Comparator 2000+ system
Catalog # LOVI460241
Supplier: Lovibond
CAS Number:
Specifications
- Chemická metoda:Silicomolybdate
- Bal.:100 mL
- Popis:Silica reagent, ammonium molybate liquid solution
- Rozsah:2.5 – 25 mg/l SiO₂