Reagents for hardness and calcium hardness using Lovibond® MD photometers
Dodavatel: Lovibond
Certifikáty
Certifikáty
O tomto produktu
Frequently Bought Together
pH buffers, ready to use
Dodavatel: WTW
pH buffers, DuraCal
Dodavatel: HAMILTON BONADUZ
Density standard, water
Dodavatel: Mettler - Toledo
VWR®, Dip Slides
Dodavatel: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Dodavatel: Merck
Pufr pH 7,00, dávkovací lahev 1 * 500 mL
Dodavatel: HAMILTON BONADUZ
Fenolftalein 1% in ethanol 70% pH-indicator
Dodavatel: VWR Chemicals
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Dodavatel: VWR Chemicals
Humidity standards
Dodavatel: ROTRONIC INSTRUMENTS
Reagents for chlorine determination using Lovibond® MD photometers
Dodavatel: Lovibond
VWR®, Zkumavky kultivační se šroubovacím uzávěrem
Dodavatel: VWR Collection
Reagents for iron determination using Lovibond® MD Photometers
Dodavatel: Lovibond
Flasks, iodine without collar, with hollow glass stopper
Dodavatel: Lenz Laborglas GmbH & CO.KG
Reagents for Phosphate Determination using Lovibond® MD photometers
Dodavatel: Lovibond