Orders LinkContactUsLinkComponent
Dosing spoons, SteriWare®
Dosing spoons, SteriWare®
Dodavatel:   Sampling Systems
Dosing spoons, SteriWare®
Dodavatel:  Sampling Systems
Certifikáty
Dosing spoons, SteriWare®
Dosing spoons, SteriWare®
2 Dostupných variant níže
Product Details & Documents
These disposable and lightweight PS spoons are ideal for sampling and dosing small amounts of powder and liquid. Ideal for work in quality control, laboratories and production.
  • Made from white shatterproof high impact polystyrene (FDA and EU 10/2011 acceptable grade)
  • Handy design comes with a 2,5 ml and 5 ml spoon on either end
  • Available in both sterile and non sterile versions
  • Cross contamination is eliminated - the device can be used straight from the bag
  • Moulded and packed in a cleanroom to assure cleanliness and hygiene
  • BSE/TSE-free
Balení: Individually bagged and shipped with full documentation including certificates of conformity and sterilisation certificates.
Product Family Options
Informace o produktu
  • Sekvence
    Kapacita
    Dostupnost
    Price
  • SAMP8642A-01
    Non sterile
    Non sterile
    01
    2,5/5,0 ml
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 130 KS
    1 * 130 KS - 1 450,00 Kč
    1 450,00 Kč
    1 * 130 KS
       1 450,00 Kč
  • Sterile
    02
    2,5/5,0 ml
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 130 KS
    1 * 130 KS - 2 020,00 Kč
    2 020,00 Kč
    1 * 130 KS
       2 020,00 Kč