Serum pipettes
18, Serum pipettes, graduated, 2,5 ml, 153 mm, -, 500/inner pack, 1500 pieces/carton
Katalogové číslo 612-1021
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
These pipettes are suitable for the rapid transfer of liquids.
Specifikace
- Balené:500/inner pack, 1500 pieces/carton
- Sterilní:-
- Kapacita:2,5 ml
- Popis:Serum pipettes, graduated
- Délka:153 mm
- Sekvence:18