Reference materials, metals in biodiesel Standard, VHG
Catalog # PROOVHG-B100M5-5
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:100 g
- Popis:Metals (Ca, K, Mg, Na, P at 5 µg/g) in B100 biodiesel
Specifications
About this item
This solution is intended for use as a calibration or reference standard for the analysis of these analytes in biodiesel or related organic matrices according to ASTM D6751. Sulfur is present in the raw materials and is not certified.
Certifications: Traceable to NIST, SRM.