Flask, Digesdahl®
458-0048
: Hach
- Popis:Flask, flat-bottom
- Bal.:1
Specially designed glass flask for use with Digesdahl® digestion apparatus. Each flask is calibrated individually with a 100 ml (to contain) mark.
Specially designed glass flask for use with Digesdahl® digestion apparatus. Each flask is calibrated individually with a 100 ml (to contain) mark.