Standards for water pollution QC check, mercury, VHG
Catalog # PROOVHG-QWPHG-15
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:15 mL
- Popis:Water analysis standard for mercury, 15 ml
- Typ:Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to mercury.
This standard has been preserved with approximately 1% (v/v) nitric acid and 0,04% (w/v) potassium dichromate. This standard has been prepared as a concentrate and must be diluted prior to analysis.
Certifications: Traceable to NIST SRM.