Catalase (from Micrococcus lysodeikticus)
SIAL60634-500ML
: Merck
:
- Bal.:500 mL
- Čí. CAS:9001-05-2
- Zdroj:Micrococcus lysodeikticus
- Sekvence:02
- Podmínky uskladnění:2...8 °C
- Formulář:Dark brown Solution
- Název enzymu:Catalase
- Aktivita enzymu:65000 - 150000 U/ml
- Molekulární hmotnost:230000
Catalase catalyzes the decomposition of hydrogen peroxide into water and oxygen. The enzyme activity will be in the range of 65000 to150000 U/mL at pH 7.0 and 25 °C.