Pipette and tip storage station, SP Bel-Art
Catalog # BELAF189619166
Supplier: Bel-Art Products, a Part of SP
Specifications
- Barva:Bílá
- Popis:Pipette stand for 6 pipettes, with tip compartments
- Š×H×V:240×420×165 mm
- Bal.:1
Specifications
About this item
This handy station keeps pipettors and tips within reach.
- Top rack stores up to six pipettes; three bins hold a generous supply of bulk pipette tips
- White acrylic plastic base, sides and top afford easy cleaning
- Bin front and self-closing lid are made from transparent acrylic for clear view of contents