Bovine Ribonuclease A (from Pancreas)
Supplier: Merck
Certificates
About this item
Ribonuclease A, type XII-A, from bovine pancreas.
- RNase protection assays
- Remove unspecifically bound RNA
- Analysis of RNA sequences
- Hydrolyze RNA contained in protein samples
- Purification of DNA
Packaging: Packed in poly bottle (10 mg) and other volumes in glass bottle.
Specifications
- Zdroj:Pancreas
- Druhy:Bovine
- Název enzymu:Ribonuclease A