Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy pro ropný průmysl
Reference standard, water saturated octanol, NIST
Reference standard, water saturated octanol, NIST
Reference standard, water saturated octanol, NIST
Catalog # PROONIST-2890
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Reference standard, water saturated octanol, NIST
Catalog # PROONIST-2890
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Bal.:
5
Popis:
NIST-2890, water saturated octanol
Specifications
About this item
Product Details
Product Details