Elemental analysis standards, portland cement type Ⅲ, NIST-1887B
PROONIST-1887B
:
- Typ balení:Vial
- Bal.:5
- Popis:NIST-1887B, Type Ⅲ portland cement
- Pk Info:5× 4 g
These are intended primarily for the evaluation and calibration of methods for analysis of cements and materials of similar matrix.
A unit consists of five vials, each containing at least 4 g of portland cement ground to pass through a 75 µm (No. 200) sieve, and each sealed in a foil pouch.
: Traceable to NIST and SRM.