Ice scraper, neoScrape
Ice scraper for cleaning freezing units, blade width 95 mm, 210 mm
Katalogové číslo KOEH2-7632
Dodavatel: KOEHLER TECHNISCHE PRODUKTEN
Přejít na:
Product Details & Documents
Special ice scraper for the professional cleaning of freezer units.
- Enables users to easily and quickly remove stubborn icing in freezers
- Blade is made from PS
- With hook for hanging
Specifikace
- Popis:Ice scraper for cleaning freezing units, blade width 95 mm
- Délka:210 mm