Pipette stand, Poxygrid®, SP Bel-Art
Catalog # BELAH189620000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Určeno pro:10 standard pipettes (Finnpette®, Socorex®, Eppendorf® pipettes); 2 pipettes with profile like Drummond Pipette-Aid®
- Barva:Modrá
- Sekvence:01
- Š×H×V:229×127×171 mm
- Bal.:1
Specifications
About this item
This epoxy-coated aluminum rack stores pipettes and protects pipette tips.
- Holds up to 10 round shape pipettors such as Finnpipette®, Socorex®, Eppendorf® and other popular brands; two end holes fit Pipette-Aid® pipette fillers
- Two stepped rows of 5 each
- Second lower support row has smaller holes to accept tapered ends of the pipettors to keep them vertical
- Quick and easy access for more efficient work; no more searching, fumbling or breakage