Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Těsnění
Filmy těsnící
Overlaminate for padlock labels
Overlaminate for padlock labels
Overlaminate for padlock labels
Catalog # BRDY065508
Supplier:
Brady
Expand image
Overlaminate for padlock labels
Catalog # BRDY065508
Supplier:
Brady
Specifications
Popis:
Clear overlaminate for padlock labels
Sekvence:
901
Tloušťka:
48 µm
Bal.:
40
Specifications
About this item
Product Details
Documentation
Product Details
Documentation