Orders LinkContactUsLinkComponent
Plug lockout, Stopower™
Plug lockout, Stopower™
Plug lockout, Červený, 27,5×18,7×44,7 mm
Katalogové číslo BRDY065673
Dodavatel:   Brady
Plug lockout, Stopower™
Katalogové číslo BRDY065673
Dodavatel:  Brady
Plug lockout, Stopower™
Plug lockout, Stopower™
Plug lockout, Červený, 27,5×18,7×44,7 mm
Přejít na:
Price
792,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
This plug hugger is ideal for office equipment and other frequently locked out cords.
  • For use on 2 or 3 pronged polarised plugs
  • Cord cannot be plugged in with Stopower in place
  • Device cannot be removed without a key
Specifikace
  • Type:
    Plug lockout
  • Barva:
    Červený
  • Š×H×V:
    27,5×18,7×44,7 mm