ContactUsLinkComponent
Human recombinant D-Amino-Acid Oxidase (from E. coli)
Human recombinant D-Amino-Acid Oxidase (from E. coli)
Catalog # PRSI92-027
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant D-Amino-Acid Oxidase (from E. coli)
Catalog # PRSI92-027
Supplier:  ProSci Inc.
CAS Number:  
Specifications
  • Bal.:
    50 µG
  • Typ enzymu:
    Recombinant
  • Zdroj:
    E. coli
  • Druhy:
    Human
  • Sekvence značek:
    N-6 His tag
  • Podmínky uskladnění:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Název enzymu:
    D-Amino-Acid Oxidase
  • Čistota:
    >95% as determined by reducing SDS-PAGE
  • Molekulární hmotnost:
    41.64 kD
  • Sekvence:
    Met1-Leu347
  • Formulace:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 1 mM DTT, 2 mM EDTA, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Testované aplikace:
    Bioassay
Specifications
About this item
D-Amino-Acid Oxidase (DAO) belongs to the DAMOX/DASOX family. DAO is a peroxisomal enzyme which founctions as a homodimer to oxidises D-amino acids to the corresponding imino acids, producing ammonia and hydrogen peroxide. D-amino-acid oxidase regulates the level of the neuromodulator D-serine in the brain, has a high activity towards D-DOPA and contributes to dopamine synthesis. D-amino-acid oxidase could act as a detoxifying agent which removes D-amino acids accumulated during aging. It also acts on a variety of D-amino acids with a preference for those having small hydrophobic side chains followed by those bearing polar, aromatic, and basic groups.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.