ContactUsLinkComponent
Human recombinant ribonuclease K6 (from cells)
  PRSI91-756
 :  
Human recombinant ribonuclease K6 (from cells)
  PRSI91-756
 :  
  • Bal.:
    50 µG
  • Typ enzymu:
    Recombinant
  • Zdroj:
    Cells
  • Druhy:
    Human
  • Sekvence značek:
    C-6 His tag
  • Podmínky uskladnění:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Název enzymu:
    Ribonuclease K6
  • Čistota:
    >95% as determined by reducing SDS-PAGE
  • Molekulární hmotnost:
    15.69 kD
  • Sekvence:
    Trp24-Leu150
  • Formulace:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mMDTT, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Testované aplikace:
    Bioassay
Ribonuclease K6 (RNASE6) is a secreted protein that belongs to the pancreatic ribonuclease family. Human RNASE6 is synthesised as a 150 amino acid precursor that contains a 23 amino acid signal sequence, and a 127 amino acid mature chain. RNASE6 is expressed in many tissues, with high expression levels in the lung, with lower expression levels in the heart, placenta, kidney, pancreas, liver, brain, and skeletal muscle. It is also detected in monocytes and neutrophils. RNASE6 may have a role in host defense.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.