Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Mouse recombinant CD47

Mouse recombinant CD47

50 µG
Catalog # PRSI92-654
Supplier:   ProSci Inc.
Mouse recombinant CD47
Catalog # PRSI92-654
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant CD47

50 µG

CAS Number:  
Price
10 420,00 Kč
Log in for availability
1 * 50 µG
Product Details & Documents

CD47 is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix. The protein is also a receptor for the C-terminal cell-binding domain of thrombospondin, and it may play a role in membrane transport and signal transduction. This protein has broad tissue distribution, and is reduced in expression on Rh erythrocytes.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Bal.:
    50 µG
  • Cat. No.:
    PRSI92-654
  • Molekulární hmotnost:
    42.8 kD
  • Čistota:
    > 95% as determined by reducing SDS-PAGE
  • Sekvence:
    Gln19-Pro158
  • Podmínky uskladnění:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Název proteinu/peptidu:
    CD47
  • Formulace:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Druhy:
    Mouse
  • Sekvence značek:
    C-Fc tag
  • Proteinová synonyma:
    IAP|MER6|Antigenic Surface Determinant Protein OA3|Leukocyte Surface Antigen CD47|Protein MER6|OA3|Integrin-Associated Protein|CD47
  • Typ proteinu/peptidu:
    Recombinant

Specifications