Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant MEK-binding 1 (from E. coli)
Human recombinant MEK-binding 1 (from E. coli)
Catalog # PRSI91-987
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant MEK-binding 1 (from E. coli)
Catalog # PRSI91-987
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Bal.:
    50 µG
  • Typ proteinu/peptidu:
    Recombinant
  • Zdroj:
    E. coli
  • Druhy:
    Human
  • Sekvence značek:
    N-6 His tag
  • Podmínky uskladnění:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Proteinová synonyma:
    MEK-Binding Partner 1|Late Endosomal/Lysosomal Adaptor and MAPK and MTOR Activator 3|MAP2K1IP1|Mp1|Mitogen-Activated Protein Kinase Scaffold Protein 1|Mitogen-Activated Protein Kinase Kinase 1-Interacting Protein 1|Ragulator Complex Protein LAMTOR3|MAPKSP1|LAMTOR3
  • Název proteinu/peptidu:
    MEK-binding 1
  • Čistota:
    > 95% as determined by reducing SDS-PAGE
  • Molekulární hmotnost:
    15.78 kD
  • Sekvence:
    Met1-Val124
  • Formulace:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 5mM DTT, 30% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Testované aplikace:
    Bioassay

Specifications

About this item

Mitogen-Activated Protein Kinase Scaffold Protein 1 (MAPKSP1) was identified as an interacting protein that belongs to the LAMTOR3 family. MAPKSP1 restricted to late endosomes by the mitogen-activated protein-binding protein-interacting protein, and binds specifically to MAP kinase kinase MAP2K1/MEK1, MAP kinase MAPK3/ERK1, and MAP kinase MAPK1/ERK2. MAPKSP1 interacts with MAP2K1/MEK1 and MAPK2 and enhances the activation of MAPK2, and thus is thought to function as an adaptor to enhance the efficiency of the MAP kinase cascade.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag