ContactUsLinkComponent
Boric acid isotopic standard, NIST-951A
  PROONIST-951A
 :  
Boric acid isotopic standard, NIST-951A
  PROONIST-951A
 :  
  • Bal.:
    10 g
  • Popis:
    Boric acid - boron isotopes 10B and 11B
  • Typ:
    Reference standard
This standard reference material is intended for use as an isotopic reference material for the calibration of mass spectrometers. The material consists of highly purified boric acid of high homogeneity.
Mixture of approx 20% Boron 10 and 80% Boron 11.
 : Traceable to NIST and SRM.