Organic reference standard, 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol dihydrochloride
Catalog # PROOLGCFOR0380.11
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:10 mg
- Standard:Reference standard
- Čí. CAS:164726-80-1
- Popis:4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol dihydrochloride
- Vzorec:C₁₉H₂₅Cl₃N₂O
- Popis:4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol dihydrochloride