Compact circuit breaker lockout
Catalog # BRDY836597
Supplier: Brady
Specifications
- Type:Circuit breaker lockout
- Barva:Červený
- Bal.:1
Specifications
About this item
Compact universal design to lock out a large range of single, double and triple-pole circuit breakers.
Place lockout over circuit breaker handle, tighten set screw and attach padlock and tag.