Detergent, disinfectant, RELY+ ON™ VIRKON®
Katalogové číslo PHAG60009
Dodavatel: PHAGOGENE
Specifikace
- Popis:RELY+ ON™ VIRKON® disinfectant
- Sekvence:03
- Balené:5 kg canister
- Bal.:5 kg
Specifikace
O tomto produktu
Disinfectant for floors, surfaces and equipment in hazardous environment.
- High oxidising power for high level disinfecting efficiency
- For wet wiping with pre-labeled spray or washing
- Colour indication to confirm the validity of the product
Certifications: Bactericide: EN 1040, EN 1276 in dirt conditions; Fungicide: EN 1275 (Candida albicans), EN 1650; Virucide: NFT 72-180.
Caution: Use biocides safely. Always read the label and product information before use.