Anti-IL17F Rabbit Polyclonal Antibody
BSBTA02062-3
- Druh protilátky:Primární
- Symbol antigenu:IL17F
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Reaktivita:Rat,Mouse
- Rozměry protilátky:100 µg
- Bal.:100 µG
Rabbit IgG polyclonal antibody for IL17F detection. Tested with WB, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat