Anti-HSPB6 Rabbit Polyclonal Antibody
BSBTA07981-1
- Druh protilátky:Primární
- Název antigenu:Heat shock protein beta-6
- Symbol antigenu:HSPB6
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Rabbit IgG polyclonal antibody for Heat shock protein beta-6(HSPB6) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: HSPB6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat