Absorbent lab mat, Chemoprotect® prep
Katalogové číslo CODA790303
Dodavatel: CODAN
Specifikace
- Balené:Sheet
- Barva:Modrá
- D׊:540×410 mm
- Bal.:50
Specifikace
O tomto produktu
The sterile Chemoprotect® prep mat has been specially developed for cytotoxic drug preparation. The threelayer structure of the material allows safe working and risk-free subsequent disposal. The prep mat is available sterile or non-sterile in two sizes for most workplaces.
- Lint-free fluid-permeable surface
- Fluid-retaining, highly absorbent middle layer
- Fluid-impermeable bottom layer