Mussel tissue - organics, NIST-2974A standard
Catalog # PROONIST-2974A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:5 g
- Standard:Reference Standard
- Popis:NIST-2974A, mussel tissue - organic
Specifications
About this item
This standard reference material is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, polybrominated diphenyl ether (PBDE) congeners, hexabromocyclododecane (HBCD) isomers, methylmercury, inorganic mercury and total mercury in marine bivalve mollusk tissue and similar matrices.
A unit consists of one bottle containing approximately 5 g of freeze-dried mussel tissue.
Certifications: Traceable to NIST and SRM numbers.