Organic reference standard, fortified human milk - organic contaminants, NIST SRM 1954
PROONIST-1954
:
- Bal.:5
- Standard:Reference standard
- Popis:Fortified human milk
This reference standard intended for use in evaluating analytical methods for the determination of selected polychlorinated biphenyl (PCB) congeners, chlorinated pesticides and polybrominated diphenyl ether (PBDE) congeners in human milk and similar matrices.
A unit consists of five bottles of approximately 5 ml fortified human milk.
: Traceable to NIST and SRM numbers.