VWR® Cell Chambers
Dodavatel: VWR Collection
O tomto produktu
VWR® cell chambers, made from medical grade, general purpose polystyrene (GPPS) and with vented cap, are ideal for large-scale cell culture.
- Available in 1-, 2-, 5- and 10-tray versions for easy scale-up
- Surface treated for optimal cell attachment and growth
- Hydrophobic membrane (pore size of 0,22 µm) avoids potential contamination during gas exchange
- Does not absorb cytokines and growth factors, and cell harvesting by conventional trypsin digestion method is possible
- Sterilised by irradiation
- Free from DNase and RNase, non pyrogenic
4 Dostupných variant níže
Product Details & Documents
VWR® cell chambers, made from medical grade, general purpose polystyrene (GPPS) and with vented cap, are ideal for large-scale cell culture.
- Available in 1-, 2-, 5- and 10-tray versions for easy scale-up
- Surface treated for optimal cell attachment and growth
- Hydrophobic membrane (pore size of 0,22 µm) avoids potential contamination during gas exchange
- Does not absorb cytokines and growth factors, and cell harvesting by conventional trypsin digestion method is possible
- Sterilised by irradiation
- Free from DNase and RNase, non pyrogenic
Documents
Avantor Cell Culture Solutions.pdf VWR for Cell Culture
Produktová literatura
CoC VWR Cell Chamber Tissue Culture Treated Vent Cap Sterile CoC_VWRI734-2980_210325530 CoC_VWRI734-2980_220514530A COC_VWRI734-2980_221009530F COC_VWRI734-2980_221109530F
More
CoC VWR Cell Chambers.pdf
Prohlášení o shodě
TDS VWR® Cell Chamber.pdf
Technický list
Specifications for Product Family
Specifikace
- Catalog No:
- 734-2980
- 734-2981
- 734-2982
- 734-3703
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifikace
Product Family Options
Informace o produktu
- SekvenceTypDoporučený pracovní objemPlocha růstuBalenéDostupnostPrice
- Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.734-2980ModelEnvironmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.734-2981ModelEnvironmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.734-2982ModelEnvironmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifikace:New Product
More
Specifications
Cat. No.734-3703ModelEnvironmentally PreferableCertifikát ochrany životního prostředíNew Product