Organic reference standards, desipramine hydrochoride standard, BP
Catalog # PROOBP931
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Reference standard
- Čí. CAS:50-47-5
- Popis:Desipramine hydrochoride
- Vzorec:C₁₈H₂₃ClN₂
- Popis:Desipramine hydrochloride