Organic reference standards, diclofenac, BP
Catalog # PROOBP828
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:25 mg
- Standard:Reference standard
- Čí. CAS:15362-40-0
- Popis:Diclofenac impurity A standard
- Vzorec:C₁₄H₉Cl₂NO
- Popis:Diclofenac impurity A