Organic reference standards, doxycycline hyclate standard, BP
Catalog # PROOBP780
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Reference standard
- Čí. CAS:24390-14-5
- Popis:Doxycycline hyclate standard
- Vzorec:C₄₆H₅₈Cl₂N₄O₁₈
- Popis:Doxycycline Hyclate