0,00 Kč
Zadání objednávky
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standardy
Standardy organické
Organic reference standards, Caffeine assay standard, BP
Organic reference standards, Caffeine assay standard, BP
PROOBP766
:
LGC Standards PROMOCHEM
:
Organic reference standards, Caffeine assay standard, BP
PROOBP766
:
LGC Standards PROMOCHEM
:
Bal.:
100 mg
Standard:
Reference standard
Čí. CAS:
58-08-2
Popis:
Caffeine assay standards
Vzorec:
C₈H₁₀N₄O₂
Popis:
Caffeine assay standard