Organic reference standards, Mercaptopurine standard, BP
Catalog # PROOBP762
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:25 mg
- Standard:Reference standard
- Čí. CAS:50-44-2
- Popis:Mercaptopurine standard
- Vzorec:C₅H₄N₄S
- Popis:Mercaptopurine assay standard