Organic reference standards, desogestrel assay standard, BP
Catalog # PROOBP568
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Reference standard
- Čí. CAS:54024-22-5
- Popis:Desogestrel assay standard
- Vzorec:C₂₂H₃₀O
- Popis:Desogestrel assay standard