Organic reference standards, spironolactone, BP
Catalog # PROOBP306
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:150 mg
- Standard:Reference standard
- Čí. CAS:52-01-7
- Popis:Spironolactone
- Vzorec:C₂₄H₃₂O₄S
- Popis:Spironolactone