Organic reference standards, megestrol acetate assay standard, BP
Catalog # PROOBP223
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Reference standard
- Čí. CAS:3562-63-8
- Popis:Megestrol acetate assay standard
- Vzorec:C₂₂H₃₀O₃
- Popis:Megestrol acetate assay standard