Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standards, lincomycin hydrochloride assay standard, BP
Organic reference standards, lincomycin hydrochloride assay standard, BP
Catalog # PROOBP215
CAS Number:  
Organic reference standards, lincomycin hydrochloride assay standard, BP
Catalog # PROOBP215
CAS Number:  

Specifications

  • Bal.:
    250 mg
  • Standard:
    Reference standard
  • Čí. CAS:
    7179-49-9
  • Popis:
    Lincomycin hydrochloride assay standard
  • Vzorec:
    C₁₈H₃₅ClN₂O₆S
  • Popis:
    Lincomycin hydrochloride assay standard

Specifications

About this item