Draining racks
With 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm), 420×170×610 mm
Katalogové číslo 149-1351
Dodavatel: WDF - Funke GmbH & Co. KG
Přejít na:
Product Details & Documents
Wire, PE-coated.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
It is designed with free-standing or for wall-mounting and with drip pan. The feet are protected by plastic caps.
Specifikace
- Popis:With 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)
- Š×H×V:420×170×610 mm