Orders LinkContactUsLinkComponent
Conductivity/ATC electrodes, 2-cell, Eutech
 :   EUTECH
Conductivity/ATC electrodes, 2-cell, Eutech
 :  EUTECH
Conductivity/ATC electrodes, 2-cell, Eutech
Conductivity/ATC electrodes, 2-cell, Eutech
Laboratory electrodes for measuring conductivity and temperature.
  • Stainless steel rings, PP and ultem-body
  • 6-pin, 8 pin or BNC connector
  • For use with different Eutech meters
Product Family Options
  • Sekvence
    Popis
    Vodivost
    Konstanta cely
    Tepl.
    Ø×D
    Připojení
    Určeno pro
  • 01
    Cela vodivostní ATC
    0 - 150 mS
    1,0 cm⁻¹
    0...80 °C
    16×144 mm
    6kolíkový, 1m kabel
    CON 510 / CON 400 /CON 410 / PC 300 / PC 510 / CON 11 / CON 110 metry
    1 * 1 KS
    1 * 1 KS - 6 660,00 Kč
    6 660,00 Kč
    1 * 1 KS
       6 660,00 Kč
  • 02
    Conductivity/temperature
    0 - 350 mS
    0,53 cm⁻¹
    0...100 °C
    12×120 mm
    8-pin, 1 m cable
    CON 2700/PC 2700
    1 * 1 KS
    1 * 1 KS - 13 200,00 Kč
    13 200,00 Kč
    1 * 1 KS
       13 200,00 Kč
  • 03
    Conductivity/ATC cell
    0 - 150 mS
    1,0 cm⁻¹
    0...80 °C
    16×144 mm
    BNC & phono plug, 1 m cable
    COND 6+ / TDS 6+ / Salt 6+ meters
    1 * 1 KS
    1 * 1 KS - 6 660,00 Kč
    6 660,00 Kč
    1 * 1 KS
       6 660,00 Kč