Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Držáky strážních sloupů
Vydac, guard cartridge holder for HPLC columns
Vydac, guard cartridge holder for HPLC columns
Vydac, guard cartridge holder for HPLC columns
GCH10
:
Avantor
Vydac, guard cartridge holder for HPLC columns
GCH10
:
Avantor
Popis:
Guard cartridge holder for 10 mm ID columns
Sekvence:
01
Bal.:
1