Scalpel handle, Cygnetic®
Catalog # SWAN5398
Supplier: SWANN MORTON
Specifications
- Popis:Handle, stainless steel, non sterile, individually sleeved
- Délka:124 mm
- Bal.:1
Specifications
About this item
This stainless steel handle is designed to assist the OR nurse with safe and easy blade attachment and removal before, during and after the procedure.
- Safe and easy blade attachment and removal
- Simple disassembly for cleaning
- Great strength and stability
- Handle is non sterile, blades are sterile
Ideal for use within rigorous Orthopaedic and general surgical procedures.
Packaging: Handle is supplied individually sleeved. Blades (to be ordered separately) are supplied individually V.P.I. protected in metal foil packets.