Test kits, photometric, Surfactants, anionic, LCK cuvette test with reagents
Catalog # 705-3068
Supplier: Hach
CAS Number:
Specifications
- Chemická metoda:Methylene Blue (MBA)
- Bal.:25 Tests
- Popis:Surfactants (anionic)
- Sekvence:273
- Typ:LCK332
- Pro jednotku:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Rozsah:0,05 - 2,0 mg/l
Specifications
About this item
Ready for use, photometric evaluation system and environmentally friendly.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Certifications: ISO 7875-1-2-1984, DIN 38409-H 23-1