Test kits, photometric, Chromic acid, LCK cuvette tests
Catalog # HACHLCK213
Supplier: Hach
CAS Number:
Specifications
- Chemická metoda:Intrinsic colour of electroplating baths
- Bal.:25 Tests
- Popis:Chromic acid
- Sekvence:214
- Typ:LCK213
- Pro jednotku:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Rozsah:0,5 - 5,0 g/l CrO₃
50 - 450 g/l CrO₃
Specifications
About this item
Pre-dosed reagents for maximum safety.
Storage conditions: 15 to 25 °C