Organic reference standards, cellulose acetate phthalate, Ph. Eur.
Catalog # EDQMY0001054
Supplier: EDQM
CAS Number:
Specifications
- Bal.:10 mg
- Standard:Reference standard
- Čí. CAS:9004-38-0
- Popis:Cellulose acetate phthalate, Ph. Eur. standard
- Vzorec:C₁₁₆H₁₁₆O₆₄
- Popis:Cellulose acetate phthalate