Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CASP1 Rabbit Polyclonal Antibody
Anti-CASP1 Rabbit Polyclonal Antibody
  BIRBORB159333-100
 :  Biorbyt
Anti-CASP1 Rabbit Polyclonal Antibody
  BIRBORB159333-100
 :  Biorbyt

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Cleaved-Caspase 1 (D210)
  • Klonovalita:
    Polyclonal
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Skladovací teplota:
    Store at –20 °C for a year
  • Imunogen:
    Synthesized peptide derived from the Internal region of human Caspase-1.
  • Bal.:
    100 µG

 

 

Type: Primary
Antigen: Cleaved-Caspase 1 (D210)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human