Orders LinkContactUsLinkComponent
BCR® flat mop systems
BCR® flat mop systems
Dodavatel:   BERKSHIRE
BCR® flat mop systems
Dodavatel:  BERKSHIRE
Certifikáty
BCR® flat mop systems
2 Dostupných variant níže
Product Details & Documents
Efficient cleaning of floors, walls and other large surfaces is critical to the overall cleanliness of the production environment. BCR® Mop Systems replicate the most effective way to clean a surface – wiping. This mop system is easy to use, highly effective and quick. The system is very flexible, combining a wide range of alternative mop wipes designed for particular levels of cleanliness with a range of flat mop heads.
  • High quality anodised aluminum
  • Can be autoclaved for use in sterile applications
  • Unique stainless steel spring mechanisms for wiper attachment
  • Plastic eyes hold mop wipers in place (not autoclavable)
Certifications: ISO Class 3–7 / Floors, walls and ceilings
Product Family Options
Flat mop heads
Informace o produktu
  • Rozměry
    Dostupnost
    Price
  • 141-1701
    Flat mop head 3
    Flat mop head 3
    90×280 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 4 800,00 Kč
    4 800,00 Kč
    1 * 1 KS
       4 800,00 Kč
Flat mop handle
Informace o produktu
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Barva
    Dostupnost
    Price
  • MicroSeal SuperSorb® wipes, ultrasonically sealed edge
    MicroSeal SuperSorb® wipes, ultrasonically sealed edge
    BERKMSSS.0909.8
    01
    Bílá
    Přihlásit se pro dostupnost
    1 * 800 KS - 46 280,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Balené
    Dostupnost
    Price
  • SatPax® 670 with 50% IPA / 50% DIW
    SatPax® 670 with 50% IPA / 50% DIW
    141-0113
    04
    25 per bag, 12 packs
    Přihlásit se pro dostupnost
    1 * 300 KS - 20 980,00 Kč